Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopa Decarboxylase/DDC Antibody, R&D Systems™

Catalog No. PPS065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS065 Supplier R&D Systems Supplier No. PPS065
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dopa Decarboxylase/DDC Polyclonal specifically detects Dopa Decarboxylase/DDC in Human, Rat, Bovine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot.

Specifications

Antigen Dopa Decarboxylase/DDC
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias AADC, AADCaromatic-L-amino-acid decarboxylase, DDC, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28
Gene Symbols DDC
Host Species Sheep
Immunogen Dopa Decarboxylase/DDC
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1644
Test Specificity Human, bovine, canine, guinea pig, rabbit, rat, and sheep ∼55 kDa DOPA Decarboxylase (DDC) in Western blots.
Target Species Human, Rat, Bovine, Canine, Guinea Pig, Rabbit, Sheep
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.