Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine beta-Hydroxylase Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $646.00

Specifications

Antigen Dopamine beta-Hydroxylase
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP255677
SDP
View Documents
Novus Biologicals
NBP255677
100 μL
Each for $646.00
Only null left
Add to Cart
 
NB403903
SDP
View Documents
Novus Biologicals
NBP25567725UL
25 μL
Each for $382.00
Only null left
Add to Cart
 
Description

Description

Dopamine beta-Hydroxylase Polyclonal specifically detects Dopamine beta-Hydroxylase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Dopamine beta-Hydroxylase
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
DBM, dopamine beta-hydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, EC 1.14.17.1
DBH
IgG
Affinity Purified
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Cancer, Lipid and Metabolism, Neuroscience
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
1621
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VQRTPEGLTLLFKRPFGTCDPKDYLIEDGTVHLVYGILEEPFRSLEAINGSGLQMGLQRVQLLKPNIPEPELPSDACTMEVQAPNIQIPSQETTYWCYIKELPKGFSRHHIIKYEPIVTKGN
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.