Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Dopamine D1R/DRD1 Antibody (887438), mFluor Violet 610 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8276MFV610
Description
Dopamine D1R/DRD1 Monoclonal antibody specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| Dopamine D1R/DRD1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 887438 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
| NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1, Met1-Thr446, Accession # P21728 | |
| 0.1 mL | |
| GPCR, Neuroscience, Neurotransmission | |
| 1812 | |
| Store at 4C in the dark. | |
| IgG2a | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            