Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D1R/DRD1 Antibody (SG2-D1a), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$207.00 - $512.00

Specifications

Antigen Dopamine D1R/DRD1
Clone SG2-D1a
Concentration 1.0 mg/mL
Dilution Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31462765), Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11060017T
SDP
View Documents
Novus Biologicals
NB11060017SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11060017
SDP
View Documents
Novus Biologicals
NB11060017
0.1 mL
Each for $512.00
Only null left
Add to Cart
 
Description

Description

Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications

Specifications

Dopamine D1R/DRD1
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating)
Unconjugated
Mouse
GPCR, Neuroscience, Neurotransmission
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A
DRD1
IgG2b κ
Protein A purified
This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor.
SG2-D1a
Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31462765), Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating
Monoclonal
Purified
RUO
Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative)
1812
Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.