Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D1R/DRD1 Antibody (SG2-D1a), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060017FR
Description
Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
Dopamine D1R/DRD1 | |
Monoclonal | |
DyLight 680 | |
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor. | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
SG2-D1a | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating | |
DRD1 | |
Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901] | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
1812 | |
Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction