Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D2 Receptor, Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9914 Supplier Bioss Supplier No. BS20730R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.

Specifications

Antigen Dopamine D2 Receptor,
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene DRD2
Gene Accession No. P14416
Gene Alias D(2) dopamine receptor; D2 receptor; D2DR; D2R; Dopamine 2 receptor; Dopamine D2 receptor; dopamine receptor 2; dopamine receptor D2; dopamine receptor D2 isoform; Drd2; Drd-2; seven transmembrane helix receptor
Gene Symbols DRD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Dopamine D2 Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1813
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.