Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D2R/DRD2 Antibody (HL1584) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325444
Description
Dopamine D2R/DRD2 Monoclonal antibody specifically detects Dopamine D2R/DRD2 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Dopamine D2R/DRD2 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Feline | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
HL1584 | |
Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:1000 | |
D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, dopamine receptor D2 isoform, seven transmembrane helix receptor | |
Recombinant protein encompassing a sequence within the N-terminus region of human Dopamine D2R/DRD2. The exact sequence is proprietary. | |
100 μL | |
GPCR, Neuroscience | |
1813 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction