Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D3R/DRD3 Antibody (5B10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315187
Description
Dopamine D3R/DRD3 Monoclonal antibody specifically detects Dopamine D3R/DRD3 in Human, Rat samples. It is validated for Western Blot, ELISASpecifications
Dopamine D3R/DRD3 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA | |
5B10 | |
Western Blot 1:500-1:5000, ELISA | |
D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205 | |
A synthesized peptide derived from human Dopamine D3R/DRD3. UniProt ID (P35462). | |
100 μg | |
GPCR | |
1814 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction