Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D3R/DRD3 Antibody (SR1747), Novus Biologicals™
SDP

Catalog No. NB259792 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259792 100 μg
NB259796 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259792 Supplier Novus Biologicals Supplier No. NBP321963100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Dopamine D3R/DRD3 Monoclonal antibody specifically detects Dopamine D3R/DRD3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Dopamine D3R/DRD3
Applications Western Blot
Classification Monoclonal
Clone SR1747
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205
Host Species Rabbit
Immunogen A synthesized peptide derived from human Dopamine D3R/DRD3 (Uniprot #: P35462)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1814
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.