Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D5 Receptor Rabbit anti-Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206717
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6717 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6717 Supplier Fabgennix Inc Supplier No. D5R501AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dopamine D5 Receptor Polyclonal antibody specifically detects Dopamine D5 Receptor in Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Dopamine D5 Receptor
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DRD5
Gene Accession No. P25115, Q8BLD9
Gene Alias D(1B) dopamine receptor; D(5) dopamine receptor; D1B; D1b receptor; D1beta dopamine receptor; D5R; DBDR; Dopamine 5 receptor; dopamine D5 receptor; dopamine receptor 5; dopamine receptor D1B; dopamine receptor D5; DRD 1B; DRD 5; Drd1b; DRD1L2; Drd5; Drd-5; Gpcr1; Hd5dr; MGC10601
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 428-478 on mouseDopamine Receptor D5 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13492, 25195
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.