Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D5R/DRD5 Antibody (889022) [Janelia Fluor™ 549], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB82861I
Description
Dopamine D5R/DRD5 Monoclonal antibody specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Dopamine D5R/DRD5 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
889022 | |
Flow Cytometry, CyTOF-ready | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5, Met1-His477, Accession # P21918 | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
1816 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction