Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D5R/DRD5 Antibody (889022), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB82861MFV500
Description
Dopamine D5R/DRD5 Monoclonal antibody specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Dopamine D5R/DRD5 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 889022 | |
| Flow Cytometry, CyTOF-ready | |
| D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
| NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5, Met1-His477, Accession # P21918 | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 1816 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction