Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D5R/DRD5 Antibody (SG4-D1b) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280701
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Dopamine D5R/DRD5 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence : 1:200-1:1000 | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Rat. Reactivity with mouse has had mixed results. It does not react with human protein. | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SG4-D1b | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
DRD5 | |
Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-Prot# P25115] | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
1816 | |
Mouse, Rat, Human (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction