Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D5R/DRD5 Antibody (SG4-D1b), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.00

Specifications

Antigen Dopamine D5R/DRD5
Clone SG4-D1b
Concentration 1 mg/mL
Dilution Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11060019T
SDP
View Documents
Novus Biologicals
NB11060019SS
0.025 mL
Each for $206.00
Only null left
Add to Cart
 
NB11060019
SDP
View Documents
Novus Biologicals
NB11060019
0.1 mL N/A N/A N/A
Description

Description

Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Dopamine D5R/DRD5
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601
DRD5
IgG1 κ
Protein A purified
Rat. Reactivity with mouse has had mixed results. It does not react with human protein.
SG4-D1b
Western Blot 1:200-1:500, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Monoclonal
Purified
RUO
Mouse, Rat, Human (Negative)
1816
Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-Prot# P25115]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.