Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine D5R/DRD5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | Dopamine D5R/DRD5 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Dopamine D5R/DRD5 Polyclonal specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Western Blot.Specifications
Dopamine D5R/DRD5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
PBS buffer, 2% sucrose | |
1816 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601 | |
The immunogen is a synthetic peptide directed towards the middle region of human Dopamine D5R/DRD5 (NP_000789.1). | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title