Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine hydrochloride, 99%

Catalog No. AAA1113614
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol

A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.

Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 243°C to 250°C
Assay Percent Range 99%
Quantity 25 g
Beilstein 3656720
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,3421
Solubility Information Soluble in water,methanol and dimethyl sulfoxide. Insoluble in ether,petroleum ether,chloroform,benzene and toluene.
Formula Weight 189.64
Percent Purity 99%
Chemical Name or Material Dopamine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-527-8
RTECSNumber UX1092000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.