Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine Receptor D4 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206712
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6712 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6712 Supplier Fabgennix Inc Supplier No. D4R401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dopamine Receptor D4 Polyclonal antibody specifically detects Dopamine Receptor D4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Dopamine Receptor D4
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DRD4
Gene Accession No. P21917, P30729, P51436
Gene Alias AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique sequence on Dopamine Receptor D4 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13491, 1815, 25432
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.