Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dopamine Transporter Monoclonal Antibody (6V-23-23)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518079
Description
This antibody does not cross-react with human samples.
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
Specifications
Dopamine Transporter | |
Monoclonal | |
1.62 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2 | |
P23977, Q01959, Q61327 | |
Slc6a3 | |
Synthetic amino acid N-terminal peptide of rat dopamine transporter. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
6V-23-23 | |
Unconjugated | |
Slc6a3 | |
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
Mouse | |
Protein A | |
RUO | |
13162, 24898, 6531 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction