Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dopamine Transporter Monoclonal Antibody (CL3123)
GREENER_CHOICE

Catalog No. PIMA535010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535010 Supplier Invitrogen™ Supplier No. MA535010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: LHYLFSSFTT ELPWIHCNNS WNSPNCSDAH PGDSSGDSSG LNDTFGTTPA AEYFERGVLH LHQSHGIDDL G Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 85%.

The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine Transporter
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3123
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc6a3
Gene Accession No. P23977, Q01959, Q61327
Gene Alias DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Gene Symbols Slc6a3
Host Species Mouse
Immunogen Recombinant protein fragment of Human SLC6A3 (Product #RP-90672)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13162, 24898, 6531
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.