Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dopamine Transporter Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA184886

Catalog No. PA184886


Only null left
Add to Cart

Description

Description

PA1-84886 detects a band at ~88 kDa in human caudate cell lysates. Rabbit anti Human dopamine transporter antibody recognizes an epitope within the C-terminal region of dopamine transporter (DAT), also known as solute carrier family 6.

The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dopamine Transporter
Polyclonal
0.01M HEPES with 0.01% BSA, 50% glycerol, 0.15M NaCl and 0.09% sodium azide; pH 7.5
Q01959, Q61327
Slc6a3
Synthetic peptide corresponding to the c-terminus region of human dopamine transporter conjugated to KLH.
100 μL
Primary
Human, Mouse, Monkey
Antibody
IgG
Immunohistochemistry (Frozen), Western Blot
Unconjugated
Slc6a3
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Rabbit
Affinity chromatography
RUO
13162, 6531
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.