Learn More
Invitrogen™ Dopamine Transporter Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA14657
Catalog No. PA14657
Only null left
Add to Cart
Description
Description
This antibody is specific for the ~88 kDa DAT protein in Western blots of SDS-solubilized human striatal samples and in IHC applications with formaldehyde-fixed human and monkey brain sections. Store at -20ºC short term, 80ºC long term.
The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dopamine Transporter
Polyclonal
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Q01959
Slc6a3
Synthetic peptide corresponding to amino acid residues from the extracellular loop 2 region of human SLC6A3 conjugated to KLH.
100 μL
Primary
Human, Monkey
Antibody
IgG
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
Slc6a3
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3
Rabbit
Affinity Chromatography
RUO
6531
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.