Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOPEY1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555723
Description
Immunogen sequence: KLETDCEHVQ PPQWLQTLMN ACSQASDFSV QSVAISLVMD LVGLTQSVAM VTGENINSVE PAQPLSPNQG RVAVVIRPPL TQGNL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 92%.
May be involved in protein traffic between late Golgi and early endosomes. [UniProt].
Specifications
DOPEY1 | |
Polyclonal | |
Unconjugated | |
DOP1A | |
B130005I07Rik; BC035275; C130028L17; D9Ertd809e; dJ202D23.2; DOP1; DOP1A; dopey family member 1; DOPEY1; homolog of yeast DOP1; KIAA1117; Protein dopey-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JWR5 | |
DOP1A | |
Recombinant protein corresponding to Human DOPEY1. Recombinant protein control fragment (Product #RP-94641). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction