Learn More
Invitrogen™ DOPEY2 Polyclonal Antibody

Description
DOPEY2 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | DOPEY2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DOP1B |
| Gene Accession No. | Q3UHQ6, Q9Y3R5 |
| Gene Alias | 0610038M01Rik; 21orf5; 2610510B01Rik; AI503246; C21orf5; Dop1b; dopey family member 2; Dopey2; homolog of yeast DOP1; KIAA0933; mKIAA0933; Protein dopey-2 |
| Gene Symbols | DOP1B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.