Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOPEY2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567223
Description
Immunogen sequence: PANLRNARNAI LEELPRTVNT MALLWNVLRK EETQKRPVDL LGATKGSSSV YFKTTKTIRQ KILDFLNPLT AHLGVQLTAA VAAVWSRKKA QRHSKMKI Highest antigen sequence identity to the following orthologs - mouse 87%, rat 86%.
May be involved in protein traffic between late Golgi and early endosomes.
Specifications
DOPEY2 | |
Polyclonal | |
Unconjugated | |
DOP1B | |
0610038M01Rik; 21orf5; 2610510B01Rik; AI503246; C21orf5; Dop1b; dopey family member 2; Dopey2; homolog of yeast DOP1; KIAA0933; mKIAA0933; Protein dopey-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3R5 | |
DOP1B | |
Recombinant Human DOPEY2. Recombinant protein control fragment (Product #RP-107860). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction