Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DORFIN Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171860UV
Description
DORFIN Polyclonal antibody specifically detects DORFIN in Human samples. It is validated for Western BlotSpecifications
DORFIN | |
Polyclonal | |
Western Blot | |
DKFZp566B1346, dorfin, Double ring-finger protein, E3 ubiquitin-protein ligase RNF19A, EC 6.3.2, EC 6.3.2.-, p38, protein p38 interacting with transcription factor Sp1, ring finger protein 19, RING finger protein 19 isoform, ring finger protein 19Ap38 protein, ring-IBR-ring domain containing protein Dorfin, RNF19 | |
The immunogen for this product maps to a region between residue 600 and 650 of human Double Ring-Finger Protein using the numbering given in entry NP_056250.3 (GeneID 25897). | |
0.1 mL | |
Zinc Finger | |
25897 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction