Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DORFIN/RNF19 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11778R

Description
This gene encodes a member of the ring between ring fingers (RBR) protein family, and the encoded protein contains two RING-finger motifs and an in between RING fingers motif. This protein is an E3 ubiquitin ligase that is localized to Lewy bodies, and ubiquitylates synphilin-1, which is an interacting protein of alpha synuclein in neurons. The encoded protein may be involved in amyotrophic lateral sclerosis and Parkinson′s disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].Specifications
DORFIN/RNF19 | |
Polyclonal | |
Unconjugated | |
RNF19A | |
AA032313; Dorfin; Double ring-finger protein; E3 ubiquitin-protein ligase RNF19A; Gametogenesis-expressed protein GEG-154; Geg-154; p38; protein p38 interacting with transcription factor Sp1; ring finger protein (C3HC4 type) 19; ring finger protein 19; RING finger protein 19A; ring finger protein 19A, E3 ubiquitin protein ligase; ring finger protein 19A, RBR E3 ubiquitin protein ligase; ring-IBR-ring domain containing protein Dorfin; Rnf19; Rnf19a; Ubce7ip2; UBCM4-interacting protein 117; ubiquitin conjugating enzyme 7 interacting protein 2; UIP117; XY body protein; Xybp | |
Rabbit | |
Protein A | |
RUO | |
25897 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NV58 | |
RNF19A | |
KLH conjugated synthetic peptide derived from human DORFIN/RNF19. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction