Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doripenem Monohydrate 98.0+%, TCI America™
SDP

Catalog No. D5249250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5249250MG 250 mg
D524950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5249250MG Supplier TCI America Supplier No. D5249250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 364622-82-2
Molecular Formula C15H26N4O7S2
Molecular Weight (g/mol) 438.514
MDL Number MFCD09749887
InChI Key NTUBEBXBDGKBTJ-WGLOMNHJSA-N
PubChem CID 636377
IUPAC Name (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-3-[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
SMILES CC1C2C(C(=O)N2C(=C1SC3CC(NC3)CNS(=O)(=O)N)C(=O)O)C(C)O.O

Specifications

Melting Point 173°C
Quantity 250 mg
Formula Weight 420.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Doripenem Monohydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.