Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.4L

Catalog No. p-7070394 Shop All Brady Corporation Products
Click to view available options
Material:
Polycoated Tagstock
Pressure Sensitive Vinyl

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in two different materials Polycoated Tagstock, Pressure Sensitive Vinyl

Specifications

Adhesive Type Acrylic, Permanent, Cold Temperature
Color Black, Orange
Material Pressure Sensitive Vinyl
Product Type Vehicle Placard
Display Text EXPLOSIVE 1.1L
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.004 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.