Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOT1L Antibody (OTI1D8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272297F
Description
DOT1L Monoclonal specifically detects DOT1L in Human samples. It is validated for Immunohistochemistry.Specifications
DOT1L | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
DOT1L | |
Recombinant protein fragment corresponding to amino acids 1-434 of human DOT1L (NP_115871) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI1D8 | |
Immunohistochemistry | |
DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4 | |
Mouse | |
Affinity Purified | |
Chromatin Research, Stem Cell Markers | |
84444.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction