Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DOT1L, Mouse, Unlabeled, Clone: 1A11.2,

Catalog No. MABE425MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE425MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE425MI Supplier MilliporeSigma™ Supplier No. MABE425
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histone-lysine N-methyltransferase, H3 lysine-79 specific (EC 2.1.1.43; UniProt Q8TEK3; also known as Disruptor of telomeric silencing-1 like, DOT1-like protein, H3-K79-HMTase, Histone H3-K79 methyltransferase, Lysine N-methyltransferase 4) is encoded by the DOT1L (also known as KIAA1814, KMT4) gene (Gene ID 84444) in human. Disruptor of telomeric silencing-1 like (DOT1L) catalyzes the mono-, di-, and tri-methylation of histone H3 on lysine-79 (H3K79). DOT1L-mediated H3K79 methylation is associated with active transcription. Accumulating evidences indicate a role of DOT1L and H3K79 in many biological processes, including DNA damage response, cell cycle, embryonic cell development, as well as the initiation and maintenance of mixed lineage leukemia (MLL)-rearranged leukemia. MLL fusion proteins such as AF9, AF10, and ENL recruit DOT1L to their target gene loci to help promote the transcription of genes that contribute to leukemic transformation of hematopoietic progenitor cells. DOT1L also has been identified as a coactivator of Wnt signaling in colorectal cancer, and is shown to induce neoplastic transformation of immortalized human breast epithelial cells and increase tumor initiation and growth of human breast cancer cells. In addtion, DOT1L is reported to regulate androgen receptor activity via direct methylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOT1L
Applications Western Blot
Classification Monoclonal
Clone 1A11.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DOT1L, KIAA1814, KMT4
Gene Accession No. Q8TEK3
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human DOT1L.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_115871
Test Specificity Clone 1A11.2 detects both spliced isoforms of human DOT1L reported by UniProt (Q8TEK3).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.