Learn More
DOT1L Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300954A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
Histone methyltransferase. Methylates ′Lys-79′ of histone H3. Nucleosomes are preferred as substrate compared to free histones. Binds to DNA.Specifications
DOT1L | |
Polyclonal | |
Unconjugated | |
DOT1L | |
KMT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84444 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8TEK3 | |
DOT1L | |
Between 1485 and C-term | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.