Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns

Catalog No. 03255024
Change view
Click to view available options
Length (Metric):
150 mm
500 mm
750 mm
Product Line:
Double nanoViper
Single nanoViper
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Product Line
03-255-024 150 mm Double nanoViper
03-255-025 500 mm Double nanoViper
03-255-026 750 mm Double nanoViper
SNV150150PN 150 mm Single nanoViper
DNV150150PN 150 mm Double nanoViper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-255-024 Supplier Thermo Scientific™ Supplier No. DNV75150PN
Add to Cart
Add to Cart

Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.

Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — Double nanoViper™ columns connect with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, Double nanoViper™
Diameter (Metric) 75 μm
Length (Metric) 150 mm
Max. Pressure 22,000 psi (1500 bar)
Particle Size 2 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m2/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Double nanoViper
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Description Double nanoViper™ PepMap™ Neo 2 μm C18 75 μm X 150 mm
For Use With (Application) Peptide Mapping, LC-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.