Learn More
Invitrogen™ Double Stranded DNA Monoclonal Antibody (AE-2)

Description
AE-2 recognizes double stranded DNA expressed in the nucleus of eukaryotes. Dilution advice:.
Specifications
Specifications
| Antigen | Double Stranded DNA |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AE-2 |
| Concentration | 0.1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | dsdna |
| Host Species | Mouse |
| Immunogen | Nuclei of Raji Burkitt's lymphoma cells. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.