Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Double stranded DNA Mouse anti-Human, Clone: DSD/958, Abnova™

Catalog No. 89034718 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-718 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-718 Supplier Abnova Corporation Supplier No. MAB14635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human double stranded DNA.

  • High specificity
  • High reactivity

Specifications

Antigen Double stranded DNA
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone DSD/958
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human double stranded DNA.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunocytochemistry (0.5-1 ug/mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determin
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Host Species Mouse
Immunogen Nuclei of Burkitt's cells.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the double stranded DNA in human cells.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG3 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.