Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

double-stranded RNA Monoclonal Antibody (J2), Exalpha
SDP

Catalog No. 502551848
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-1848 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-1848 Supplier Vector Laboratories Supplier No. 10010200200UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nucleic acid separation prior to dsRNA-immunoblotting must be carried out by polyacrylamide gel electrophoresis, because the sensitivity of detection is considerably lower after blotting from agarose gels.

double-stranded RNA Monoclonal antibody specifically detects double-stranded RNA in Yeast samples. It is validated for Affinity Purification, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen double-stranded RNA
Applications Affinity Purification, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone J2
Conjugate Unconjugated
Description 100UG HUMAN CD68 KP1 AFP555 MAB 100 UG
Formulation PBS with no preservative
Host Species Mouse
Immunogen mixture of 50 μg L-dsRNA and 75 μg methylated bovine serum albumin
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.