Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Antibody (3E1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192684IR
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Doublecortin | |
Monoclonal | |
DyLight 755 | |
O43602 | |
DCX | |
Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Cow, Pig and other Mammals. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
30 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
Mouse | |
49.3 kDa | |
0.1 mL | |
Neuroscience, Phospho Specific, Signal Transduction | |
1641 | |
Human, Mouse, Rat, Pig, Bovine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction