Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Antibody (3E1), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | Doublecortin |
---|---|
Clone | 30 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP192684SS
![]() |
Novus Biologicals
NBP192684SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP192684
![]() |
Novus Biologicals
NBP192684 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Doublecortin | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Neuroscience, Signal Transduction | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
DCX | |
IgG2a | |
Protein G purified | |
Human, Mouse, Rat, Cow, Pig and other Mammals. |
30 | |
Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bovine, Equine | |
O43602 | |
1641 | |
Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
35-45 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title