Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Antibody (OTI4A3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270558MFV500
Description
Doublecortin Monoclonal antibody specifically detects Doublecortin in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Doublecortin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI4A3 | |
Flow Cytometry | |
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
Full length human recombinant protein of human DCX (NP_835365) produced in HEK293T cell. | |
0.1 mL | |
Neuroscience, Phospho Specific, Signal Transduction | |
1641 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction