Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doublecortin Antibody (OTI4A3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270558PE
Description
Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Doublecortin | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
DCX | |
Full length recombinant protein of human Doublecortin (NP_835365) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI4A3 | |
Flow Cytometry | |
doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin) | |
Mouse | |
Affinity Purified | |
Neuroscience, Phospho Specific, Signal Transduction | |
1641.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction