Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Doublecortin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517428 Supplier Invitrogen™ Supplier No. PA517428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Doublecortin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 165 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene DCX
Gene Accession No. O43602, O88809, Q9ESI7
Gene Alias DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS
Gene Symbols DCX
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human doublecortin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13193, 1641, 84394
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.