Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X2, 100-200 mesh, ion-exchange resin

Catalog No. p-2885569
Click to view available options
Quantity:
500 g
2.5 kg

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

Physical Form Beads
Particle Size Particle size analysis: on 50 mesh (cumulative): < 15%, through 140 mesh: < 15%
Packaging Plastic Bottle
Volume Total Capacity wet volume capacity, Cl form (meq/mL):> 0.6
Water Retention Capacity Cl form: 70 to 80%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Merck Index 10, 2182
Quantity 500 g
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Dowex™ 1X2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.