Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X8, 50-100 mesh, ion-exchange resin

Catalog No. p-2885571
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Dowex 1X8, C29H34ClN, CAS Number-12627-85-9, dowex 1x8 50-100 cl, dowex 1x2 50-100 cl, 500g, White, 01,511; 04,267; 06,302; 07,182; 08,263, BBQMUEOYPPPODD-UHFFFAOYSA-M, 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride, MFCD00132718, 432.048, Plastic bottle | CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES *

Specifications

Physical Form Powder and/or Chunks
Particle Size 15 % max. (on 35 mesh, cumulative) 15 % max. (through 80 mesh)
Packaging Plastic bottle
Volume Total Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 43 to 48%
Color White
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Quantity 2.5 kg
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
Chemical Name or Material Dowex™ 1X8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.