Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 1X8-50, ion-exchange resin, Thermo Scientific™

Catalog No. AC202981000
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69772-06-1

Specifications

Physical Form Beads
Particle Size Particle size analysis: on 35 mesh (cumulative): <15%, through 80 mesh: <15%
Volume Total Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 43 to 48%
Color Amber to White
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Merck Index 10, 2182
Quantity 100 g
Chemical Name or Material Dowex(r) 1X8-50
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.