Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50WX8, 100-200 mesh, ion-exchange resin

Catalog No. p-2885576
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle

CAS: 11119-67-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Physical Form Moist Beads or Spheres
Particle Size 15% Max. (on 60 mesh, cumulative) 15 % Max. (through 170 mesh)
Packaging Plastic bottle
Volume Total Capacity wet volume capacity, H form min. (meq/mL): 1.7
Water Retention Capacity H form: 50 to 58%
Color Beige to Brown
Quantity 2.5 kg
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Dowex™ 50WX8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.