Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowex™ 50WX4, 200-400 mesh, ion-exchange resin

Catalog No. p-2885581
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle

CAS: 11113-61-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11113-61-4
MDL Number MFCD00132725

Specifications

Physical Form Beads
Particle Size Particle size analysis: on 100 mesh (cumulative): <15%, through 325 mesh: <15%
Packaging Plastic bottle
Volume Total Capacity wet volume capacity, H form (meq/mL):> 1.1
Water Retention Capacity H form: 64 to 72%
Quantity 2.5 kg
Chemical Name or Material Dowex™ 50WX4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.