Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec Down Draft Work Cell

Shop All Mopec Products


Click to view available options
Height (Metric):
20.32cm
Description
- Designed to fit in a 30 in. front to back countertop
- Removable work surface provides easy cleaning
- Down draft work cell has a 3.5 in. cup drain for any overflow and wash downs
- Using downdraft fume exhaust technology the BF600 down draft work cell provides a safe economical down draft working environment
- Ideal for working with reagents, formalin, xylene and alcohol
- Requires a sink base cabinet
Warranty and Services
1 Year Warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Includes | 3.5 in. (8.89cm) Cup drain for any overflow |
| Ventilation | Down draft |
| Height (English) | 8 in. |
| Width (English) | 68.58 in. |
| Height (Metric) | 20.32cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction