Learn More
Mystaire™ Downflow Workstation Main Filter
Click to view available options
For Use With (Application):
For Formaldehyde- FE-WS6
For Hydrocarbons- FE-WS6
For Mineral Acids- FE-WS6
Description
- Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Specifications
For Use With (Equipment) | FE-WS6 |
Material | Carbon |
For Use With (Application) | For Formaldehyde- FE-WS6 |
Type | Filter |