Learn More
Dowtherm™ A, Thermo Scientific Chemicals
Description
A fluid may be used in systems employing either liquid phase or vapor phase heating. Suitable applications include indirect heat transfer. Mainly in oil & gas, plastic processing, chemical processing, solar energy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A fluid may be used in systems employing either liquid phase or vapor phase heating. Suitable applications include indirect heat transfer. Mainly in oil & gas, plastic processing, chemical processing, solar energy.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 12°C to 14°C |
Boiling Point | 257°C |
Odor | Unpleasant |
UN Number | UN3082 |
Density | 1.056 |
Quantity | 1000 mL |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Dowtherm™ A |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
EINECSNumber | 616-827-0 |
RTECSNumber | DV1500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.