Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dowtherm™ A

Catalog No. AAH303470F
Change view
Click to view available options
Quantity:
1000 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH303470F 2500 mL
AAH30347AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH303470F Supplier Thermo Scientific Chemicals Supplier No. H303470F
Only null left
Add to Cart

CAS: 8004-13-5 | C24H20O | 324.423 g/mol

A fluid may be used in systems employing either liquid phase or vapor phase heating. Suitable applications include indirect heat transfer. Mainly in oil & gas, plastic processing, chemical processing, solar energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluid may be used in systems employing either liquid phase or vapor phase heating. Suitable applications include indirect heat transfer. Mainly in oil & gas, plastic processing, chemical processing, solar energy.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-13-5
Molecular Formula C24H20O
Molecular Weight (g/mol) 324.423
MDL Number MFCD00148859
InChI Key MHCVCKDNQYMGEX-UHFFFAOYSA-N
Synonym diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
PubChem CID 24670
IUPAC Name 1,1'-biphenyl;phenoxybenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2

Specifications

Melting Point 12°C to 14°C
Boiling Point 257°C
Odor Unpleasant
UN Number UN3082
Density 1.056
Quantity 2500 mL
Solubility Information Insoluble in water.
Chemical Name or Material Dowtherm™ A
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 616-827-0
RTECSNumber DV1500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.