Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxepin Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602326
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail

D1195, 1229-29-4, C19H21NO·HCl

Spectrum™ Chemical Doxepin Hydrochloride, USP is a tricyclic antidepressant used to treat mental and mood problems such as depression and anxiety. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1229-29-4
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.84
InChI Key MHNSPTUQQIYJOT-SJDTYFKWSA-N
IUPAC Name hydrogen dimethyl({3-[(2E)-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene]propyl})amine chloride
SMILES [H+].[Cl-].CN(C)CC\C=C1/C2=CC=CC=C2COC2=CC=CC=C12

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.